Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-2 R beta Antibody (5H4), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB153729 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB153729 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB153729 Supplier Novus Biologicals Supplier No. NBP312041DL594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-2 R beta Monoclonal antibody specifically detects IL-2 R beta in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen IL-2 R beta
Applications Flow Cytometry
Classification Monoclonal
Clone 5H4
Conjugate DyLight 594
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-2 receptor subunit beta, IL-2R subunit beta, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75
Host Species Rat
Immunogen This antibody was raised by immunising Lewis rats with transfected rat myeloma cell line YB2/0, expressing truncated murine IL-2R beta.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3560
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.