Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-2 R beta Antibody (TM-b1), PE, Novus Biologicals™
SDP

Catalog No. NBP143355PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143355PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143355PE Supplier Novus Biologicals Supplier No. NBP143355PE
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-2 R beta Monoclonal specifically detects IL-2 R beta in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-2 R beta
Applications Flow Cytometry
Classification Monoclonal
Clone TM-b1
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-2 receptor subunit beta, IL-2R subunit beta, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75
Gene Symbols IL2RB
Host Species Rat
Immunogen The immunogen for this antibody was Purified CD122.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3560
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.