Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-20 R beta/FNDC6 Antibody (20RNTC), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP200466V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP200466V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP200466V Supplier Novus Biologicals Supplier No. NBP200466V
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-20 R beta/FNDC6 Monoclonal specifically detects IL-20 R beta/FNDC6 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IL-20 R beta/FNDC6
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 20RNTC
Conjugate DyLight 405
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Gene Alias FNDC6, DIRS1, IL-20R2, IL20RB interleukin 20 receptor beta, MGC34923
Gene Symbols IL20RB
Host Species Rat
Immunogen The immunogen for this antibody is FNDC6 (IL-20R2).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53833
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.