Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-20 R beta/FNDC6 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB082689 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB082689 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB082689 Supplier Novus Biologicals Supplier No. NBP298428F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-20 R beta/FNDC6 Polyclonal specifically detects IL-20 R beta/FNDC6 in Rat samples. It is validated for ELISA.

Specifications

Antigen IL-20 R beta/FNDC6
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias FNDC6, DIRS1, IL-20R2, IL20RB interleukin 20 receptor beta, MGC34923
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant IL-20 R beta/FNDC6 (B8K1X9; Met1-Pro227).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53833
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.