Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-20R alpha Antibody (173714), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB314765 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314765 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314765 Supplier Novus Biologicals Supplier No. FAB11762PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-20R alpha Monoclonal antibody specifically detects IL-20R alpha in Human samples. It is validated for Flow Cytometry

Specifications

Antigen IL-20R alpha
Applications Flow Cytometry
Classification Monoclonal
Clone 173714
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20R1IL-20 receptor subunit alpha, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-20 R alpha , Val30-Lys250, Accession # Q9UHF4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53832
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.