Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-21 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176740SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176740SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176740SS Supplier Novus Biologicals Supplier No. NBP176740SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-21 Polyclonal specifically detects IL-21 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IL-21
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Blocking Assay
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2 ug/mL, Block/Neutralize
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_068575
Gene Alias IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform
Gene Symbols IL21
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the center of human IL-21 precursor. The immunogen is located within the last 50 amino acids of IL-21. Amino Acid Squence: TCPSCDSYEKKPPKE
Molecular Weight of Antigen 17 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Research Discipline Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 59067
Test Specificity At least two isoforms of IL-21 are known to exist; this antibody will detect both isoforms. IL-21 antibody is predicted to not cross-react with other members of the interleukin protein family.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.