Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-21 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB194375 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB194375 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB194375 Supplier Novus Biologicals Supplier No. NBP176740DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-21 Polyclonal antibody specifically detects IL-21 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen IL-21
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the center of human IL-21 precursor. The immunogen is located within the last 50 amino acids of IL-21. Amino Acid Squence: TCPSCDSYEKKPPKE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 59067
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.