Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-21 Antibody (RM0268-6G53), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NBP21696646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP21696646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP21696646 Supplier Novus Biologicals Supplier No. NBP211696JF646

Rat Monoclonal Antibody

IL-21 Monoclonal specifically detects IL-21 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-21
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone RM0268-6G53
Conjugate Janelia Fluor 646
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform
Gene Symbols IL21
Host Species Rat
Immunogen Mouse recombinant IL-21
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 59067
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.