Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-21R Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB197348 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB197348 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB197348 Supplier Novus Biologicals Supplier No. NBP176739R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-21R Polyclonal antibody specifically detects IL-21R in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen IL-21R
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias CD360 antigen, IL-21 receptor, IL-21R, interleukin 21 receptor, interleukin-21 receptor, MGC10967, NILR, Novel interleukin receptor
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to amino acids 97 to 111 of human IL-21 receptor precursor. The immunogen is located within amino acids 80 - 130 of IL-21 Receptor. Amino Acid Squence: NITDQSGNYSQECGS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chemokines and Cytokines, Cytokine Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 50615
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.