Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-22 Antibody (034), DyLight 488
SDP

Catalog No. NB070477 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB070477 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB070477 Supplier Novus Biologicals Supplier No. NBP290160G

Rabbit Monoclonal Antibody

IL-22 Monoclonal specifically detects IL-22 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen IL-22
Applications ELISA
Classification Monoclonal
Clone 34
Conjugate DyLight 488
Dilution ELISA, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-22 (NP_065386.1; Ala34-Ile179).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 50616
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.