Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-22 Antibody (140301R), FITC, Novus Biologicals™
SDP

Catalog No. NB296026 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296026 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296026 Supplier Novus Biologicals Supplier No. FAB582RF
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-22 Monoclonal specifically detects IL-22 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen IL-22
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 140301R
Conjugate FITC
Dilution Intracellular Staining by Flow Cytometry
Formulation PBS
Gene Alias Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Host Species Rat
Immunogen E. coli-derived recombinant mouse IL-22, Leu34-Val179, Accession # Q9JJY9
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 50616
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.