Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-22 Antibody (8F6F10), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP227322G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227322G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227322G Supplier Novus Biologicals Supplier No. NBP227322G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-22 Monoclonal specifically detects IL-22 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-22
Applications Western Blot
Classification Monoclonal
Clone 8F6F10
Conjugate DyLight 488
Dilution Western Blot
Gene Accession No. Q9GZX6
Gene Alias Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Gene Symbols IL22
Host Species Mouse
Immunogen Full-length recombinant human IL-22 protein was used as immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 50616
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.