Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-22 Antibody (8F6F10), Novus Biologicals™
SDP

Catalog No. NBP227322SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227322SS 0.025 mg
NBP227322 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227322SS Supplier Novus Biologicals Supplier No. NBP227322SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-22 Monoclonal specifically detects IL-22 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-22
Applications Western Blot
Classification Monoclonal
Clone 8F6F10
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 2 ug/mL
Gene Accession No. Q9GZX6
Gene Alias Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Gene Symbols IL22
Host Species Mouse
Immunogen Full-length recombinant human IL-22 protein was used as immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 50616
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.