Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-22 Antibody (8F6F10), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $438.00

Specifications

Antigen IL-22
Clone 8F6F10
Concentration 0.5 mg/mL
Dilution Western Blot 0.5 - 2 ug/mL
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227322SS
SDP
View Documents
Novus Biologicals
NBP227322SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP227322
SDP
View Documents
Novus Biologicals
NBP227322
0.1 mg
Each for $438.00
Only null left
Add to Cart
 
Description

Description

IL-22 Monoclonal specifically detects IL-22 in Human samples. It is validated for Western Blot.
Specifications

Specifications

IL-22
0.5 mg/mL
Western Blot
Unconjugated
Mouse
Biologically Active Proteins, Cytokine Research
Q9GZX6
50616
Full-length recombinant human IL-22 protein was used as immunogen for this antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
8F6F10
Western Blot 0.5 - 2 ug/mL
Monoclonal
Purified
RUO
Human
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL-D110, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
IL22
IgG2b κ
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.