Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-22BP/IL22 RA2 Antibody (001), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB306377 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306377 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB306377 Supplier Novus Biologicals Supplier No. NBP289745MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-22BP/IL22 RA2 Monoclonal antibody specifically detects IL-22BP/IL22 RA2 in Human samples. It is validated for ELISA

Specifications

Antigen IL-22BP/IL22 RA2
Applications ELISA
Classification Monoclonal
Clone 001
Conjugate mFluor Violet 500 SE
Dilution Sandwich ELISA Capture
Formulation 50mM Sodium Borate
Gene Alias class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 receptor subunit alpha-2, IL-22BPCRF2X, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-22BP/IL22 RA2 (Accession#: NP_851826.1; Met1-Pro231).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 116379
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.