Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-22BP/IL22 RA2 Antibody (875504), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB339890 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339890 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339890 Supplier Novus Biologicals Supplier No. FAB10871MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-22BP/IL22 RA2 Monoclonal specifically detects IL-22BP/IL22 RA2 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining).

Specifications

Antigen IL-22BP/IL22 RA2
Applications CyTOF, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 875504
Conjugate mFluor Violet 500 SE
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 receptor subunit alpha-2, IL-22BPCRF2X, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-22BP, Thr22-Pro263, Accession # Q969J5
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 116379
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.