Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-22BP/IL22 RA2 Antibody (MM0397-6G47) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP211700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211700 0.1 mg
NB018859 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP211700 Supplier Novus Biologicals Supplier No. NBP211700

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

IL-22BP/IL22 RA2 Monoclonal specifically detects IL-22BP/IL22 RA2 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-22BP/IL22 RA2
Applications Western Blot
Classification Monoclonal
Clone MM0397-6G47
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
Gene Accession No. NP_851826
Gene Alias class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 receptor subunit alpha-2, IL-22BPCRF2X, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16
Gene Symbols IL22RA2
Host Species Mouse
Immunogen Human recombinant IL-22BP/IL22 RA2
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 116379
Reconstitution Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.