Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-22BP/IL22 RA2 Antibody (MM0397-6G47) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
Antigen | IL-22BP/IL22 RA2 |
---|---|
Clone | MM0397-6G47 |
Concentration | LYOPH |
Applications | Western Blot |
Classification | Monoclonal |
Description
IL-22BP/IL22 RA2 Monoclonal specifically detects IL-22BP/IL22 RA2 in Human samples. It is validated for Western Blot.Specifications
IL-22BP/IL22 RA2 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Human | |
class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 receptor subunit alpha-2, IL-22BPCRF2X, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16 | |
IL22RA2 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
MM0397-6G47 | |
Western Blot | |
Unconjugated | |
Mouse | |
Cytokine Research, Immunology, Signal Transduction | |
NP_851826 | |
116379 | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title