Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-22BP/IL22 RA2 Goat anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1007420.025MG
Description
IL-22BP/IL22 RA2 Polyclonal specifically detects IL-22BP/IL22 RA2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IL-22BP/IL22 RA2 | |
Polyclonal | |
Unconjugated | |
10mM KHPO4 and 0.14M NaCl, pH 7.2 with 0.1% Sodium Azide | |
class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 receptor subunit alpha-2, IL-22BPCRF2X, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
IL22 RA2 | |
Store at -20C. Avoid freeze-thaw cycles. |
Immunohistochemistry (Paraffin), Immunohistochemistry, ELISA | |
1.0 mg/mL | |
ELISA 1:50000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
Q969J5 | |
IL22RA2 | |
Synthetic peptide RVQFQSRNFHNILQWQPGRALTGNSSVY-C corresponding to 33-60 residues of N-terminus of human IL-22R-alpha-2 protein with cysteine added for conjugation to a carrier protein. | |
0.025 mg | |
Cytokine Research, Immunology, Signal Transduction | |
116379 | |
Mouse, Rat, Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction