Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-22BP/IL22 RA2 Goat anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody
$181.90
Specifications
Antigen | IL-22BP/IL22 RA2 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | ELISA 1:50000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 |
Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, ELISA |
Classification | Polyclonal |
Description
IL-22BP/IL22 RA2 Polyclonal specifically detects IL-22BP/IL22 RA2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IL-22BP/IL22 RA2 | |
ELISA 1:50000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
Polyclonal | |
Goat | |
Cytokine Research, Immunology, Signal Transduction | |
10mM KHPO4 and 0.14M NaCl, pH 7.2 with 0.1% Sodium Azide | |
class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 receptor subunit alpha-2, IL-22BPCRF2X, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16 | |
IL22RA2 | |
IgG | |
Affinity Purified | |
IL22 RA2 |
1.0 mg/mL | |
Immunohistochemistry (Paraffin), Immunohistochemistry, ELISA | |
Unconjugated | |
RUO | |
Mouse, Rat, Human | |
Q969J5 | |
116379 | |
Synthetic peptide RVQFQSRNFHNILQWQPGRALTGNSSVY-C corresponding to 33-60 residues of N-terminus of human IL-22R-alpha-2 protein with cysteine added for conjugation to a carrier protein. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title