Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-22R alpha 1 Antibody (496514), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB370353 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370353 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370353 Supplier Novus Biologicals Supplier No. FAB42941I

Rat Monoclonal Antibody

IL-22R alpha 1 Monoclonal antibody specifically detects IL-22R alpha 1 in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen IL-22R alpha 1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 496514
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R1, IL-22RA1, IL-22R-alpha-1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-22 R alpha 1, Thr18-Ala228, Accession # Q80XZ4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 58985
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.