Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-22R alpha 1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB194736 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB194736 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB194736 Supplier Novus Biologicals Supplier No. NBP176724AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-22R alpha 1 Polyclonal antibody specifically detects IL-22R alpha 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen IL-22R alpha 1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R1, IL-22RA1, IL-22R-alpha-1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to amino acids near the amino terminus of human IL-22 receptor precursor. The immunogen is located within the first 50 amino acids of IL-22 Receptor. Amino Acid Squence: QSSNFENILTWDSGPE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 58985
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.