Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-23A/IL-23 P19 Antibody (727753) [DyLight 550], Novus Biologicals™
SDP

Catalog No. NB399070 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399070 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399070 Supplier Novus Biologicals Supplier No. FAB17161L
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-23A/IL-23 P19 Monoclonal antibody specifically detects IL-23A/IL-23 P19 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen IL-23A/IL-23 P19
Applications Western Blot, ELISA
Classification Monoclonal
Clone 727753
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias IL-23, IL-23A, IL-23-A, IL-23p19, IL23P19P19, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-23 subunit alpha, Interleukin-23 subunit p19, JKA3 induced upon T-cell activation, MGC79388, SGRFIL-23 subunit alpha
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-23 p19, Arg20-Pro189, Accession # Q9NPF7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cytokine Research, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 51561
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.