Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-23R Antibody (15N6C6), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP227092IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227092IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227092IR Supplier Novus Biologicals Supplier No. NBP227092IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-23R Monoclonal specifically detects IL-23R in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IL-23R
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 15N6C6
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry-Paraffin
Gene Accession No. Q5VWk5
Gene Alias IBD17, IL-23 receptor, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Gene Symbols IL23R
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 288-315 of hIL-23 receptor was used as immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 149233
Test Specificity There are multiple isoforms of IL-23 receptor. Researchers are encouraged to use a tool like BLAST to determine the suitability of this antibody for their testing.
Target Species Human, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.