Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-23R Antibody (218213) [Biotin], Novus Biologicals™
SDP

Catalog No. NB402941 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402941 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402941 Supplier Novus Biologicals Supplier No. FAB14001B

Mouse Monoclonal Antibody

IL-23R Monoclonal antibody specifically detects IL-23R in Human samples. It is validated for Western Blot

Specifications

Antigen IL-23R
Applications Western Blot
Classification Monoclonal
Clone 218213
Conjugate Biotin
Dilution Western Blot
Formulation PBS
Gene Alias IBD17, IL-23 receptor, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-23 R, Gly24-Ile354, Accession # Q5VWK5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 149233
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.