Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-24 Antibody (101), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB069666 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB069666 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB069666 Supplier Novus Biologicals Supplier No. NBP290083AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IL-24 Monoclonal specifically detects IL-24 in Human samples. It is validated for ELISA.

Specifications

Antigen IL-24
Applications ELISA
Classification Monoclonal
Clone 101
Conjugate Alexa Fluor 700
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias C49A, FISP, IL10B, IL-24, interleukin 24, mda-7, MDA7MOB5, melanocyte-associated Mda-7, melanoma differentiation association protein 7, Melanoma differentiation-associated gene 7 protein, Mob-5, ST16interleukin-24, suppression of tumorigenicity 16 (melanoma differentiation), Suppression of tumorigenicity 16 protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-24 (NP_001172085.1; Met1-Leu207).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 11009
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.