Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-24 Antibody (MM0398-6F48), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP211702FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211702FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211702FR Supplier Novus Biologicals Supplier No. NBP211702FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-24 Monoclonal specifically detects IL-24 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-24
Applications Western Blot
Classification Monoclonal
Clone MM0398-6F48
Conjugate DyLight 680
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C49A, FISP, IL10B, IL-24, interleukin 24, mda-7, MDA7MOB5, melanocyte-associated Mda-7, melanoma differentiation association protein 7, Melanoma differentiation-associated gene 7 protein, Mob-5, ST16interleukin-24, suppression of tumorigenicity 16 (melanoma differentiation), Suppression of tumorigenicity 16 protein
Gene Symbols IL24
Host Species Mouse
Immunogen Human recombinant IL-24
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 11009
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.