Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-24 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056407T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056407T 0.025 mg
NB10056407 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056407T Supplier Novus Biologicals Supplier No. NB10056407SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-24 Polyclonal specifically detects IL-24 in Human, Primate samples. It is validated for Flow Cytometry, Western Blot.

Specifications

Antigen IL-24
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-3 ug/ml, Flow Cytometry 2-5 ug/10^6 cells
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Alias C49A, FISP, IL10B, IL-24, interleukin 24, mda-7, MDA7MOB5, melanocyte-associated Mda-7, melanoma differentiation association protein 7, Melanoma differentiation-associated gene 7 protein, Mob-5, ST16interleukin-24, suppression of tumorigenicity 16 (melanoma differentiation), Suppression of tumorigenicity 16 protein
Gene Symbols IL24
Host Species Rabbit
Immunogen Amino acids 123-136 (KNYHNRTVEVRTLK) of human IL-24 were used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11009
Test Specificity The amino acid sequence used as immunogen is found in many of the IL-24 isoforms, including isoforms of 219, 207, 206, 154, 143 and 120 amino acids.
Target Species Human, Primate, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.