Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-26/AK155 Antibody (510414) [DyLight 755], Novus Biologicals™
SDP

Catalog No. NB400202 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400202 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400202 Supplier Novus Biologicals Supplier No. FAB13751Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-26/AK155 Monoclonal antibody specifically detects IL-26/AK155 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen IL-26/AK155
Applications Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 510414
Conjugate DyLight 755
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias AK155IL-26interleukin-26, interleukin 26, Protein AK155
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-26/AK155, Lys22-Gln171, Accession # Q9NPH9.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 55801
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.