Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-26/AK155 Antibody (MM0399-6D20) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018861 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018861 0.025 mg
NBP211703 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018861 Supplier Novus Biologicals Supplier No. NBP2117030.025MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

IL-26/AK155 Monoclonal specifically detects IL-26/AK155 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-26/AK155
Applications Western Blot
Classification Monoclonal
Clone MM0399-6D20
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. Q9NPH9
Gene Alias AK155IL-26interleukin-26, interleukin 26, Protein AK155
Gene Symbols IL26
Host Species Mouse
Immunogen Human IL26
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 55801
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.