Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-27 Antibody (307426), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB304375 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304375 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304375 Supplier Novus Biologicals Supplier No. FAB25261S

Mouse Monoclonal Antibody

IL-27 Monoclonal antibody specifically detects IL-27 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen IL-27
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 307426
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias IL-27, IL-27 p28 subunit, IL-27 subunit alpha, IL27A, IL-27-A, IL27-A, IL27p28, IL30, interleukin 27, interleukin 30, interleukin-27 subunit alpha, MGC71873, p28IL-27A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-27, Arg21-Lys229 of EBI-3 (Accession #Q14213.2) and Phe29-Pro243 of p28 (Accession #AAM34498)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 246778
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.