Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-27 Antibody (355025), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB367417 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB367417 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB367417 Supplier Novus Biologicals Supplier No. FAB18342E

Rat Monoclonal Antibody

IL-27 Monoclonal antibody specifically detects IL-27 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen IL-27
Applications Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 355025
Conjugate DyLight 405
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias IL-27, IL-27 p28 subunit, IL-27 subunit alpha, IL27A, IL-27-A, IL27-A, IL27p28, IL30, interleukin 27, interleukin 30, interleukin-27 subunit alpha, MGC71873, p28IL-27A
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-27 heterodimer, Tyr19-Pro228 (EBI3), Phe29-Ser234 (p28), Accession # O35228 (EBI3) & Q8K3I6 (p28)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 246778
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.