Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-27 p28/IL-30 Antibody (234205), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB296979 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296979 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296979 Supplier Novus Biologicals Supplier No. FAB1834R

Rat Monoclonal Antibody

IL-27 p28/IL-30 Monoclonal antibody specifically detects IL-27 p28/IL-30 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen IL-27 p28/IL-30
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 234205
Conjugate Alexa Fluor 647
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias IL27,IL-27,IL27A,IL-27A,IL27p28,IL-27p28,IL30,p28
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse IL-27, Tyr19-Pro228 (EBI3) and Phe29-Ser234 (p28), Accession # O35228 (EBI3) and Q8K3I6 (p28)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.