Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-27R alpha/WSX-1/TCCR Antibody (263503), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB313768 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313768 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313768 Supplier Novus Biologicals Supplier No. FAB21091APCCY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-27R alpha/WSX-1/TCCR Monoclonal antibody specifically detects IL-27R alpha/WSX-1/TCCR in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen IL-27R alpha/WSX-1/TCCR
Applications Flow Cytometry
Classification Monoclonal
Clone 263503
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL27R, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-27 R alpha/WSX-1/TCCR, Gly29-Lys510, Accession # O70394
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 9466
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.