Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-27R alpha/WSX-1/TCCR Antibody (MM0563-8A5), Biotin, Novus Biologicals™
SDP

Catalog No. NBP211917B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211917B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211917B Supplier Novus Biologicals Supplier No. NBP211917B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-27R alpha/WSX-1/TCCR Monoclonal specifically detects IL-27R alpha/WSX-1/TCCR in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-27R alpha/WSX-1/TCCR
Applications Western Blot
Classification Monoclonal
Clone MM0563-8A5
Conjugate Biotin
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL27R, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1
Gene Symbols IL27RA
Host Species Mouse
Immunogen Human recombinant IL-27R alpha/WSX-1/TCCR (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 9466
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.