Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-28A Mouse anti-Human, Clone: 3B4A1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | IL-28A |
|---|---|
| Clone | 3B4A1 |
| Concentration | 1.3 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6367
![]()
|
Proteintech Group Inc
602701IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
IL28 is a cytokine distantly related to type I interferons and the IL-10 family. This gene, interleukin 28B (IL28B), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).Specifications
| IL-28A | |
| 1.3 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Cytokine Zcyto20, IFN lambda 2, IFNL2, IL 28A, IL28A, IL-28A, Interferon lambda 2, Interleukin 28A, ZCYTO20 | |
| IFNL2 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 3B4A1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8IZJ0 | |
| 282616 | |
| IL-28A Fusion Protein Ag18525 | |
| Primary | |
| -20°C | |
| IFNL2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title