Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-28B/IFN-lambda 3 Antibody (617), Novus Biologicals™
SDP

Catalog No. NBP243738 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP243738 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP243738 Supplier Novus Biologicals Supplier No. NBP243738
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot

IL-28B/IFN-lambda 3 Monoclonal specifically detects IL-28B/IFN-lambda 3 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-28B/IFN-lambda 3
Applications Western Blot
Classification Monoclonal
Clone 617
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000
Formulation PBS with No Preservative
Gene Alias interleukin-28B, IL28B, IL-28B, IL28C, interferon, lambda 3
Gene Symbols IFNL3
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human IL28B. The exact sequence is proprietary.
Molecular Weight of Antigen 22 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 282617
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.