Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-28B/IFN-lambda 3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB396448 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396448 0.025 mg
NBP241236 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396448 Supplier Novus Biologicals Supplier No. NBP2412360.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-28B/IFN-lambda 3 Polyclonal specifically detects IL-28B/IFN-lambda 3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IL-28B/IFN-lambda 3
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% Sodium Azide
Gene Alias interleukin-28B, IL28B, IL-28B, IL28C, interferon, lambda 3
Gene Symbols IFNL3
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid peptide near the center of human IL-28B. The immunogen is located within amino acids 120 - 170 of IL-28B . Amino Acid Squence: IQPQPTAGPRTRGRLH
Molecular Weight of Antigen 16 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282617
Test Specificity IL-28B antibody is human specific. IL-28B antibody may cross-react with IL-28A and IL-29.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.