Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-28B/IFN-lambda 3 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB343669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343669 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343669 Supplier Novus Biologicals Supplier No. NBP241236MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-28B/IFN-lambda 3 Polyclonal antibody specifically detects IL-28B/IFN-lambda 3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen IL-28B/IFN-lambda 3
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias interleukin-28B, IL28B, IL-28B, IL28C, interferon, lambda 3
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid peptide near the center of human IL-28B. The immunogen is located within amino acids 120 - 170 of IL-28B . Amino Acid Squence: IQPQPTAGPRTRGRLH
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 282617
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.