Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-29/IFN-lambda 1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB362825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362825 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362825 Supplier Novus Biologicals Supplier No. NBP177893AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-29/IFN-lambda 1 Polyclonal antibody specifically detects IL-29/IFN-lambda 1 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen IL-29/IFN-lambda 1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias cytokine Zcyto21, IFNL1, IFN-lambda-1, IL-29, interferon lambda-1, interferon, lambda 1, interleukin 29 (interferon, lambda 1), interleukin-29, ZCYTO21
Host Species Rabbit
Immunogen This IgG fraction IL-29/IFN-lambda 1 Antibody was prepared from rabbit antiserum after repeated immunizations with mature length recombinant human IL-29/IFN-lambda 1/IFN-lambda 1 protein produced in E.coli. (Uniprot: Q8IU54)
Purification Method Multi-step
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 282618
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.