Learn More
Invitrogen™ IL-29 Monoclonal Antibody (247801)
Mouse Monoclonal Antibody
$536.00 - $670.50
Specifications
Antigen | IL-29 |
---|---|
Clone | 247801 |
Concentration | 0.5 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | ELISA, Neutralization |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA523791
|
Invitrogen™
MA523791 |
500 μg |
Each for $670.50
|
|
|||||
PIMA523903
|
Invitrogen™
MA523903 |
100 μg |
Each for $536.00
|
|
|||||
Description
In sandwich ELISAs, 100% cross-reactivity with recombinant human (rh) IL-28B/IFN-lambda 3 and 20% with rhIL-28A is observed. No cross-reactivity or interference was observed with rhIFN-alpha A, rhIFN-alpha B2, rhIFN-alpha G, rhIFN-alpha I, or rhIFN- beta 1A. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IFN-lambdas constitute a novel family of interferons that mediate the induction of anti-viral protection in a wide variety of cells. There are three members in this family, namely lambda1, lambda2, and lambda3, which are also known as IL-29, IL-28A, and IL-28B, respectively. Similar to type I IFNs, IFN-lambdas activate an intracellular signaling to promote gene expression. IFN-lambdas mediate their anti-viral protection through a class II cytokine receptor complex composed of two essential receptor proteins, CRF2-12/IFN-lambdaR1, which is unique to IFN-lambdas, and CFR2-4/IL-10R2, which is shared with IL-10, IL-22, and IL-26 receptors. The IFN-lambdas, IL-28 and IL-29, have recently been reported to prime dendritic cells to induce proliferation of Foxp3-bearing regulatory T cells. DCs that differentiate in response to IFN-lambdas express high levels of class I and II MHC gene products, but low levels of costimulatory molecules. Moreover, these cells can specifically induce IL-2-dependent proliferation of CD4+CD25+FOXP3+ T cells with contact-dependent suppressive activity.Specifications
IL-29 | |
0.5 mg/mL | |
ELISA, Neutralization | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 5% trehalose and No Preservative | |
Q8IU54 | |
282618 | |
Mouse myeloma cell line NS0-derived recombinant human IL-29/IFN-lambda 1 Gly20-Thr200. | |
Antibody |
247801 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Monoclonal | |
Lyophilized | |
IgG2a | |
Human | |
IFNL1 | |
cytokine Zcyto21; Cytokine Zcyto22; FNL1; IFN lambda1; IFNL1; IFNL3; IFN-lambda-1; IFN-lambda-3; IFN-lambda-4; IL28B; IL-28B; IL28C; IL-28C; IL29; IL-29; ILN; interferon lambda 1; interferon lambda 3; Interferon lambda1; interferon lambda-1; Interferon lambda-3; interferon, lambda 1; interferon, lambda 3; interferon, lambda 4; interferon-lambda-1; Interleukin; interleukin 29 (interferon, lambda 1); interleukin-28B; Interleukin-28C; interleukin-29; ZCYTO21; ZCYTO22 | |
IFNL1 | |
Primary | |
Protein A/G |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.