Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-3 Antibody (001), FITC
SDP

Catalog No. NB068871 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB068871 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB068871 Supplier Novus Biologicals Supplier No. NBP290009F

Rabbit Monoclonal Antibody

IL-3 Monoclonal specifically detects IL-3 in Human samples. It is validated for ELISA.

Specifications

Antigen IL-3
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate FITC
Dilution ELISA
Formulation PBS
Gene Alias Hematopoietic growth factor, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-3 (NP_002955.2; Met 1-Phe 152).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3562
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.