Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-3 Antibody (OTI3B11), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP271023A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271023A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271023A32 Supplier Novus Biologicals Supplier No. NBP271023AF532

Mouse Monoclonal Antibody

IL-3 Monoclonal specifically detects IL-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IL-3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3B11
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Hematopoietic growth factor, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor
Gene Symbols IL3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 20-152 of human IL-3 (NP_000579) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3562
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.