Learn More
Invitrogen™ IL-3 beta Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Goat Polyclonal Antibody
Supplier: Invitrogen™ 500P177GBT500UG

Description
AA Sequence of recombinant protein: MISDRGSDAH HLLRTLDCRT IALEILVKLP YPQVSGLNNS DDKANLRNST LRRVNLDEFL KSQEEFDSQD TTDIKSKLQK LKCCIPAAAS DSVLPGVYNK DLDDFKKKLR FYVIHLKDLQ PVSVSRPPQP TSSSDNFRPM TVEC. Preparation: Produced from sera of goats immunized with highly pure Recombinant Rat IL-3beta. Anti-Rat IL-3beta-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Rat IL-3beta by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Rat IL-3beta (500-P177G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat IL-3beta. Western Blot: To detect Rat IL-3beta by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat IL-3beta is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P177GBT-1MG will be provided as 2 x 500 μg
IL-3 is a hematopoietic growth factor that promotes the survival, differentiation and proliferation of committed progenitor cells of the megakaryocyte, granulocyte-macrophage, erythroid, eosinophil, basophil and mast cell lineages. Produced by T cells, mast cells and eosinophils, IL-3 enhances thrombopoiesis, phagocytosis, and antibody-mediated cellular cytotoxicity. Its ability to activate monocytes suggests that IL-3 may have additional immunoregulatory roles. Many of the IL-3 activities depend upon co-stimulation with other cytokines. IL-3 is a species-specific, variably glycosylated cytokine.
Specifications
IL-3 beta | |
Polyclonal | |
Biotin | |
Il3 | |
Interleukin | |
Goat | |
Antigen affinity chromatography | |
RUO | |
24495 | |
-20°C | |
Lyophilized |
ELISA, Western Blot | |
0.1-1.0 mg/mL | |
PBS with no preservative | |
P04823 | |
Il3 | |
E.coli-derived Recombinant Rat IL-3beta. | |
500 μg | |
Primary | |
Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.