Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-31 Mouse anti-Human, Clone: B7-B0, Novus Biologicals™
SDP

Catalog No. NBP266905 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266905 Supplier Novus Biologicals Supplier No. NBP266905

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

IL-31 Monoclonal specifically detects IL-31 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen IL-31
Applications ELISA, Western Blot
Classification Monoclonal
Clone B7-B0
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, ELISA 1:5000-1:10000
Formulation PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias IL-31interleukin-31, interleukin 31
Gene Symbols IL31
Host Species Mouse
Immunogen Recombinant protein within Human IL-31 aa 1-164/ 164. (SwissProt: Q6EBC2 Human)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 386653
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.