Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-31RA Antibody (313308), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB313271 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313271 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313271 Supplier Novus Biologicals Supplier No. FAB2769PEATT594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-31RA Monoclonal specifically detects IL-31RA in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-31RA
Applications Flow Cytometry
Classification Monoclonal
Clone 313308
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias class I cytokine receptor, CRL3Glmr, Cytokine receptor-like 3, GLMR, GLM-RMGC125346, GP130 like receptor, Gp130-like monocyte receptor, Gp130-like receptor, hGLM-R, IL-31 receptor subunit alpha, IL-31R subunit alpha, IL-31RAGPLCRL, IL-31R-alpha, interleukin 31 receptor A, interleukin-31 receptor subunit alpha, PRO21384, soluble type I cytokine receptor CRL3, ZcytoR17
Host Species Rat
Immunogen NS0-derived rhIL-31 RA extracellular domain, aa 20-516, Accession # NP_620586
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 133396
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.