Learn More
Description
Specifications
Specifications
| Antigen | IL-32 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 403 |
| Conjugate | DyLight 650 |
| Dilution | Sandwich ELISA Detection |
| Formulation | 50mM Sodium Borate |
| Gene Alias | IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4IL-32delta, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-32 (NP_001012651.1; Met1-Lys131). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
