Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-33 Antibody (6H617), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP227333R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227333R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227333R Supplier Novus Biologicals Supplier No. NBP227333R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-33 Monoclonal specifically detects IL-33 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen IL-33
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 6H617
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry
Gene Accession No. O95760
Gene Alias C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2
Gene Symbols IL33
Host Species Mouse
Immunogen Full length recombinant protein
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90865
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.