Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-33 Antibody (IL33305B), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB120729 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120729 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120729 Supplier Novus Biologicals Supplier No. NBP311681V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-33 Monoclonal specifically detects IL-33 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Direct ELISA.

Specifications

Antigen IL-33
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Monoclonal
Clone IL33305B
Conjugate DyLight 405
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Direct ELISA
Formulation 50mM Sodium Borate
Gene Alias C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2
Host Species Mouse
Immunogen Recombinant human IL-33.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cellular Markers, Core ESC Like Genes, Immunology, Inflammation, Innate Immunity, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 90865
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.