Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-33 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224858SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224858SS 0.025 mg
NBP224858 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224858SS Supplier Novus Biologicals Supplier No. NBP224858SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IL-33 Polyclonal specifically detects IL-33 in Human, Canine, Monkey samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen IL-33
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Simple Western 1:200
Formulation 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide
Gene Accession No. O95760
Gene Alias C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2
Gene Symbols IL33
Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human IL-33 was used as the immunogen for this antibody.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90865
Test Specificity 100% homologous in human (isoforms CRA_a and CRA_b).
Target Species Human, Canine, Monkey
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.